AC5H49B83933 Accela ChemBio Inc.

2-(4-Chlorophenyl)-2-cyclopentyl-2-hydroxyacetic Acid

  • MDL number MFCD26656977
  • Cas Number : 92021-41-5
  • Molecular Weight : 254.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY122102&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY122102.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart