AC5H49B83934 Accela ChemBio Inc.

3-Bromo-2,5-difluoropyridine

  • MDL number MFCD12827553
  • Cas Number : 1211331-43-9
  • Molecular Weight : 193.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY122294&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY122294.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart