AC5H49B8393B Accela ChemBio Inc.

2-Amino-8-nitro-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD26665235
  • Cas Number : 1369382-71-7
  • Molecular Weight : 179.1362

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY129544&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY129544.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart