AC5H49B8394B Accela ChemBio Inc.

(3aR,8aS)-3,3a,8,8a-Tetrahydro-2H-indeno[1,2-d]oxazol-2-one

  • MDL number MFCD00674088
  • Cas Number : 135969-65-2
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY223970&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY223970.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart