AC5H49B8394E Accela ChemBio Inc.

1,4-Dioxaspiro[4.5]dec-7-en-8-yl Trifluoromethanesulfonate

  • MDL number MFCD14635485
  • Cas Number : 170011-47-9
  • Molecular Weight : 288.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY225437&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY225437.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart