AC5H49B83957 Accela ChemBio Inc.

2-(6-Azaspiro[2.5]octan-6-yl)aniline

  • MDL number MFCD31716109
  • Cas Number : 2155431-25-5
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY237494&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY237494.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart