AC5H49B83959 Accela ChemBio Inc.

(3S,4S)-1-Boc-pyrrolidine-3,4-dicarboxylic Acid

  • MDL number MFCD31699921
  • Cas Number : 2170869-18-6
  • Molecular Weight : 259.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY238097&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY238097.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart