AC5H49B83968 Accela ChemBio Inc.

3-(2-Iodoethyl)oxetane

  • MDL number MFCD20486578
  • Cas Number : 1782577-96-1
  • Molecular Weight : 212.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249454&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249454.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart