AC5H49B8396C Accela ChemBio Inc.

6-Fluoro-alpha-methylpyridine-3-methanamine

  • MDL number MFCD18384299
  • Cas Number : 905587-45-3
  • Molecular Weight : 140.1581632

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249602&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249602.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart