AC5H49B8396E Accela ChemBio Inc.

2-Amino-4-methoxypyrimidine-5-boronic Acid Pinacol Ester

  • MDL number MFCD18382978
  • Cas Number : 944401-63-2
  • Molecular Weight : 251.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249630&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249630.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart