AC5H49B83973 Accela ChemBio Inc.

2-(5-Bromo-2-pyrimidinyl)-2-methylpropanal

  • MDL number MFCD31814459
  • Cas Number : 2228785-78-0
  • Molecular Weight : 229.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249673&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249673.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart