AC5H49B83977 Accela ChemBio Inc.

6-Methoxypyrazine-2-boronic Acid

  • MDL number MFCD11973849
  • Cas Number : 934346-27-7
  • Molecular Weight : 153.93168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY249976&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY249976.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart