AC5H49B8397C Accela ChemBio Inc.

8-Chloro-5-methylpyrido[2,3-b]pyrazin-6(5H)-one

  • MDL number MFCD30722506
  • Cas Number : 1613289-32-9
  • Molecular Weight : 195.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Pale yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250210&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250210.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart