AC5H49B8397D Accela ChemBio Inc.

3-Boc-6-fluoro-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD06797502
  • Cas Number : 155601-71-1
  • Molecular Weight : 237.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250431&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250431.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart