AC5H49B83980 Accela ChemBio Inc.

3-Acetyl-4-bromobenzonitrile

  • MDL number MFCD22493256
  • Cas Number : 1822653-33-7
  • Molecular Weight : 224.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250785&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250785.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart