AC5H49B83984 Accela ChemBio Inc.

5-Chloro-4-iodo-2-(methylthio)pyrimidine

  • MDL number MFCD29764933
  • Cas Number : 107403-27-0
  • Molecular Weight : 286.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250867&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250867.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart