AC5H49B83985 Accela ChemBio Inc.

4-Bromo-3-(1,1-difluoroethyl)benzonitrile

  • Molecular Weight : 246.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY250871&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY250871.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart