AC5H49B83988 Accela ChemBio Inc.

Ethyl 3-Cyano-3-phenylbutanoate

  • MDL number MFCD31562562
  • Cas Number : 123064-05-1
  • Molecular Weight : 217.2637

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251113&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251113.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart