AC5H49B8398E Accela ChemBio Inc.

4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoyl Chloride

  • MDL number MFCD07368530
  • Cas Number : 222541-76-6
  • Molecular Weight : 222.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251345&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251345.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart