AC5H49B83992 Accela ChemBio Inc.

[4-Methoxy-2-(methylthio)pyrimidin-5-yl]boronic Acid Pinacol Ester

  • MDL number MFCD16996591
  • Molecular Weight : 282.16686

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251382&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251382.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart