AC5H49B83997 Accela ChemBio Inc.

2-Oxo-2,6,7,8-tetrahydropyrrolo[1,2-a]pyrimidine-3-carboxylic Acid

  • MDL number MFCD20645691
  • Molecular Weight : 180.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251471&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251471.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart