AC5H49B8399C Accela ChemBio Inc.

7-Bromochroman-4-amine

  • MDL number MFCD09949875
  • Cas Number : 1016804-06-0
  • Molecular Weight : 228.0858

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251620&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251620.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart