AC5H49B8399F Accela ChemBio Inc.

N-(Methoxymethyl)-1,1-diphenyl-N-[(trimethylsilyl)methyl]methanamine

  • MDL number MFCD32068405
  • Cas Number : 1170701-52-6
  • Molecular Weight : 313.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY251660&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY251660.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart