AC5H49B839A2 Accela ChemBio Inc.

2-Nitro-6,7-dihydropyrazolo[1,5-a]pyridine

  • MDL number MFCD32068409
  • Cas Number : 2007081-99-2
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY252757&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY252757.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart