AC5H49B839A4 Accela ChemBio Inc.

Bis(3-fluoro-4-aminophenyl)disulfide

  • Cas Number : 16766-33-9
  • Molecular Weight : 284.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY252806&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY252806.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart