AC5H49B839A5 Accela ChemBio Inc.

3-(3,5-Difluorophenyl)acrylaldehyde

  • MDL number MFCD28992285
  • Molecular Weight : 168.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY252821&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY252821.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart