AC5H49B839A6 Accela ChemBio Inc.

5-Methoxy-3-methylbenzothiophene

  • MDL number MFCD18452494
  • Cas Number : 25784-98-9
  • Molecular Weight : 178.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY252861&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY252861.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart