AC5H49B839A7 Accela ChemBio Inc.

2-(6-Amino-3-pyridyl)-2-propanol

  • MDL number MFCD18802400
  • Cas Number : 843643-03-8
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(6-amino-3-pyridinyl)-2-propanol
Inchi Code 1S/C8H12N2O/c1-8(2,11)6-3-4-7(9)10-5-6/h3-5,11H,1-2H3,(H2,9,10)
Inchi Key BBZSDJGBSXMOON-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form Yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY252867&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY252867.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart