AC5H49B839AE Accela ChemBio Inc.

8-Bromo-6-iodoimidazo[1,2-a]pyridine

  • MDL number MFCD28099878
  • Cas Number : 1823379-07-2
  • Molecular Weight : 322.92853

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253023&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253023.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart