AC5H49B839AF Accela ChemBio Inc.

2-(4-Bromobenzyl)-1H-imidazole

  • MDL number MFCD21969106
  • Cas Number : 1368766-62-4
  • Molecular Weight : 237.09586

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253029&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253029.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart