AC5H49B839B1 Accela ChemBio Inc.

Methyl 3H-Imidazo[4,5-c]pyridine-7-carboxylate

  • MDL number MFCD32068422
  • Cas Number : 2112668-56-9
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253078&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253078.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart