AC5H49B839B4 Accela ChemBio Inc.

5-Chloro-8-iodo-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD21364530
  • Cas Number : 1030626-93-7
  • Molecular Weight : 279.46559

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253156&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253156.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart