AC5H49B839B5 Accela ChemBio Inc.

2,7-Diaminoanthraquinone

  • MDL number MFCD01664478
  • Cas Number : 605-44-7
  • Molecular Weight : 238.2414

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253157&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253157.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart