AC5H49B839B8 Accela ChemBio Inc.

3,4-Dichloropyridine-2-carboxamide

  • MDL number MFCD11877974
  • Cas Number : 1025720-99-3
  • Molecular Weight : 191.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010638&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY010638.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart