AC5H49B839B9 Accela ChemBio Inc.

2-Oxo-3-butenoic Acid

  • MDL number MFCD19231895
  • Cas Number : 56842-76-3
  • Molecular Weight : 100.07276

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253275&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253275.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart