AC5H49B839BA Accela ChemBio Inc.

3-(6-Quinolyl)propanal

  • MDL number MFCD24071407
  • Cas Number : 476660-18-1
  • Molecular Weight : 185.22184

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253547&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253547.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart