AC5H49B839BD Accela ChemBio Inc.

Ethyl 1-Amino-3-cyclopentenecarboxylate Hydrochloride

  • MDL number MFCD31617613
  • Cas Number : 76910-10-6
  • Molecular Weight : 191.65526

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253759&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253759.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart