AC5H49B839C0 Accela ChemBio Inc.

5-Bromo-2-(phenylmethylthio)pyrimidine

  • MDL number MFCD16660315
  • Cas Number : 79686-18-3
  • Molecular Weight : 281.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 5-bromo-2-pyrimidinyl sulfide
Inchi Code 1S/C11H9BrN2S/c12-10-6-13-11(14-7-10)15-8-9-4-2-1-3-5-9/h1-7H,8H2
Inchi Key LXOANVAJTYXINB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253811&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253811.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart