AC5H49B839C1 Accela ChemBio Inc.

Methyl 1-Boc-3-methyl-2-oxo-2,3-dihydro-1H-imidazole-4-carboxylate

  • MDL number MFCD32068466
  • Molecular Weight : 256.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY253812&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY253812.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart