AC5H49B839C7 Accela ChemBio Inc.

(S)-2-Methylazetidine [(1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate

  • MDL number MFCD31705062
  • Cas Number : 1877329-24-2
  • Molecular Weight : 303.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY257732&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY257732.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart