AC5H49B839CC Accela ChemBio Inc.

1-[5-(Aminomethyl)-2-pyridyl]-N,N-dimethylmethanamine

  • Cas Number : 1528646-05-0
  • Molecular Weight : 165.2355

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY261643&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY261643.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart