AC5H49B839CD Accela ChemBio Inc.

4-(tert-Butoxy)-3,3-dimethyl-4-oxobutanoic Acid

  • MDL number MFCD24496460
  • Cas Number : 660423-00-7
  • Molecular Weight : 202.24752

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY261751&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY261751.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart