AC5H5815FBA5 Accela ChemBio Inc.

3-(Boc-amino)-4-[(4-nitrobenzyl)oxy]-4-oxobutanoic Acid

  • MDL number MFCD32177090
  • Molecular Weight : 368.3386

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262124&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262124.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart