AC5H5815FBA6 Accela ChemBio Inc.

2-(3-Buten-1-yl)-4-methyl-3-nitropyridine

  • MDL number MFCD32177092
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY262178&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY262178.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart