AC5H5815FBC3 Accela ChemBio Inc.

Ethyl 1-Benzyl-3-oxopiperidine-4-carboxylate

  • MDL number MFCD00044512
  • Cas Number : 39514-19-7
  • Molecular Weight : 261.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY030648&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY030648.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart