AC5H5815FBC7 Accela ChemBio Inc.

Ethyl 7-Fluoroindole-2-carboxylate

  • MDL number MFCD16036844
  • Cas Number : 348-31-2
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY039534&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY039534.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart