AC5H5815FBC9 Accela ChemBio Inc.

Ethyl 2,2-Difluoro-2-iodoacetate

  • MDL number MFCD00798135
  • Cas Number : 7648-30-8
  • Molecular Weight : 249.9825764

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040928&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY040928.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart