AC5H5815FBCF Accela ChemBio Inc.

O6-[4-(Aminomethyl)benzyl]guanine

  • MDL number MFCD07367624
  • Cas Number : 674799-96-3
  • Molecular Weight : 270.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY047354&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY047354.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart