AC5H5815FBD0 Accela ChemBio Inc.

Dimethyl 3-Bromo-2-oxopentanedioate

  • MDL number MFCD20526357
  • Cas Number : 148728-48-7
  • Molecular Weight : 253.04736

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY047406&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY047406.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart