AC5H5815FBDC Accela ChemBio Inc.

1-Cyano-4-(dimethylamino)pyridinium Tetrafluoroborate

  • MDL number MFCD00011998
  • Cas Number : 59016-56-7
  • Molecular Weight : 234.9897128

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY058029&attname=GoodsCode

Safety Information

signalword Warning
pcodes P260-P264-P270-P280-P301+P330+P331+P310-P303+P361+P353+P310+P363-P304+P340+P310
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY058029.pdf
hstatements H302-H314
Availability details for
Quantity
  • Availability
  • Add to Cart